Whatman 6703-3610 Polycap HD PP Capsule Filter, 1.0um, 385cm2, 1/4-3/8" ID; 1/Pk

Representative image only.

Whatman 6703-3610 Polycap HD PP Capsule Filter, 1.0um, 385cm2, 1/4-3/8" ID; 1/Pk

Whatman | Mfr: 6703-3610| Item: UX-29700-20

Use polypropylene (PP) membrane filters for solvents, clean air/gas, prefiltration, and sample preparation

$106.60USD / Each

$106.60USD / Each

Buy It Together

Current Item

Whatman 6703-3610 Polycap HD PP Capsule Filter, 1.0um, 385cm2, 1/4-3/8" ID; 1/Pk
Whatman 6703-3610 Polycap HD PP Capsule Filter, 1.0um, 385cm2, 1/4-3/8" ID; 1/Pk
2970020

$106.60USD / Each
+
Whatman 6702-3600 HEPA-CAP Filter; 36 capsule, 1pk
2970092

$118.95USD / Pkg of 1
=
Total Price 225.55 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Autoclavable

More About this Item

Capsules feature sturdy PP housings with fused seals—no glues or adhesives to contaminate filtrate. Capsules include a luer lock vent to guarantee that no filtration area is wasted. Use vent to draw solution into the capsule or let air escape from the capsule or sample port.

CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.

Specifications
  • Membrane MaterialPolypropylene
  • Pore Size (µm)1
  • Effective Filter Area (cm²)385
  • Connections1/4" to 3/8" stepped barb
  • DescriptionPolycap HD PP Capsule Filter, 1.0um, 385cm2, 1/4-3/8" ID; 1/Pk
Reviews
Q&A