Cole-Parmer® ST-200 Series Ultra-Thin Magnetic Stirrer
Ultra-slim design provides convenience and saves valuable space
  • Use simple touch-and-slide bar to set stirring speed and timer
  • LCD displays real-time parameters
  • IP65-rated polypropylene housing withstands spills and dust
Ultra-flat stirrer utilizes advanced magnetic coil technology providing lighter, quieter, and wear-free operation with no moving parts. The unique design makes the stirrer thinner and lighter to be easily stored when not in use, saving valuable bench space.
Multiple stirring options allow a choice between one direction or reversing every 45 seconds for thorough mixing. Built-in timer can be set for up to 9 hours or continuous operation.
$207.75USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (in) High Speed (rpm) Power (VAC)
Availability Pricing
COMPARE
1500 100 to 240
$207.75USD / Each