Cole-Parmer® INC-250 Series Mini CO2 Digital Incubator
Space-saving footprint—ideal for use inside biological safety cabinets
  • Dual-beam infrared sensor continuously monitors CO2 levels
  • Large LED display indicate real-time temperature and CO2 percentage
  • Large 20-L internal chamber accepts bottles and flasks up to 2 L
  • Audible and visual alarms trigger when CO2 levels drop
The mini digital temperature and CO2 control incubators are only 13 inches wide, small enough to fit in most biological safety cabinets, conserving valuable bench space. A thick layer of high-efficiency insulation and a 12 VDC car adapter make this incubator ideal for transporting cells. The unit can maintain set temperature levels within 10% for up to one hour after power has been disconnected.
Intuitive control panel and large LED displays allow you to digitally adjust both temperature and CO² percentages and view in real time. Another function on the panel provides simple in-lab calibration of temperature or CO². You can add up to 80% humidity to the chamber by inserting a stainless steel reservoir.
$5,170.00 - $5,205.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Max Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
20 55 115
$5,170.00USD / Each
COMPARE
20 55 230
$5,205.00USD / Each