Cole-Parmer® OVV-400 Series Programmable Vacuum Ovens
Easily manage up to eight programs with multiple steps each
  • Stainless steel inner chamber resists corrosion
  • Rounded inner chamber corners offer easy cleaning
  • Built-in alarm alerts when oven shuts down
  • Able to heat samples up to 392°F (200°C)
These programmable vacuum ovens allow you to manage up to eight programs with eight steps each, one in which temperature, timer, and fan speed can be set. The PID temperature controller provides accurate and reliable temperature control up to 392°F (200°C). An air inlet offers easy access to inert gases such as nitrogen. The ovens feature independent vacuum interface and vents. They are also equipped with a solenoid valve to control vacuum degree and vacuum pump.
Easily monitor your samples through the protective glass window. The rubber O-ring and door lock handle provide a secure seal preventing any air leakage or unauthorized access. Additional oven features include vacuum gauge indicator, temperature deviation alarm, overtemperature set point alarm, and overcurrent protection alarm. The unit will cut off the heater automatically if the temperature is out of control.
$2,290.00 - $5,795.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Power (VAC) Rise Time
Availability Pricing
COMPARE
24 120 90 mins to 100°C
$2,290.00USD / Each
COMPARE
24 220 90 mins to 100°C
$2,290.00USD / Each
COMPARE
53 120 80 mins to 100°C
$3,625.00USD / Each
COMPARE
53 220 80 mins to 100°C
$3,625.00USD / Each
COMPARE
91 120 80 mins to 100°C
$5,795.00USD / Each
COMPARE
91 220 80 mins to 100°C
$5,795.00USD / Each
COMPARE
24 220 90 mins to 100°C
$2,290.00USD / Each
COMPARE
53 220 80 mins to 100°C
$3,625.00USD / Each
COMPARE
91 220 80 mins to 100°C
$5,795.00USD / Each
Showing 1- 9 of 9