General Tools Digital Handheld Psychrometers
Compact size allows for convenient storage
  • Small enough to fit in your pocket
Meters feature min/max memory storage, HOLD degF/degC, and auto shutoff after 20 minutes of nonuse. A large LCD simultaneoulsy displays Relative Humidity and temperature readings. Models 37952-10 and -20 display dew point. Model 37952-20 also displays wet bulb.
ITEM(S) ON SALE
$167.87USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Max RH Range (%) Min RH Range (%)
Availability Pricing
General Tools SAM990DW Digital Psychrometer, RH/, Temp, Dew Point, and Wet Bulb EW-37952-20 Mfr # SAM990DW
COMPARE
2 AAA (included) 100 0
$167.87USD / Each
$197.50USD / Each