Eagle Flammable Liquid Safety Cabinets
Adjustable steel shelves hold up to 350 lb
  • Four adjustable galvanized leveling legs
  • Trilingual warning labels
  • Raised leakproof sill
  • Meeta NFPA Code 30 and OSHA requirements
Flammable cabinets are constructed of a high gloss powder coated finish, double-walled, 18-gauge steel with 1½" (3.8 cm) insulating air barrier — ideal for safely storing flammable liquid containers up to 5 gallons in size. Cabinets feature dual vents, each have 2" (5.1 cm) threaded fittings and a fire baffle and cap, grounding attachments, red trilingual warning, 2" (5.1 cm) raised, leakproof door sill, 3-point latch system, and flush mounted door handle with key lock. A concealed self-close mechanism provides obstruction-free access to top shelf space.
Includes adjustable galvanized steel shelves which direct spills to back and bottom of leakproof sump and hold 350 lbs (158.8 kg) Uniform Distributed Load (UDL). Hazard label: FLAMMABLE-KEEP FIRE AWAY.
$918.00 - $3,832.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Number of Shelves Self-Closing
Availability Pricing
Eagle 1903X Flammable Liquid Safety Cabinet, Bench Top, Self Closing, 1 Door, 4 Gal EW-09418-20 Mfr # 1903X
COMPARE
4 1 Yes
$1,410.00USD / Each
Eagle 1904X Flammable Liquid Safety Cabinet, Bench Top, Manual Close, 1 Door, 4 Gal EW-09418-21 Mfr # 1904X
COMPARE
4 1 No
$918.00USD / Each
Eagle 1924X Flammable Liquid Safety Cabinet, Space Saver, Self Closing, 1 Door, 12 Gal EW-09418-22 Mfr # 1924X
COMPARE
12 1 Yes
$1,822.00USD / Each
Eagle 1925X Flammable Liquid Safety Cabinet, Space Saver, Manual Close, 1 Door, 12 Gal EW-09418-23 Mfr # 1925X
COMPARE
12 1 No
$1,027.00USD / Each
Eagle 3010X Flammable Liquid Safety Cabinet, Self Closing, 2 Doors, 30 Gal EW-09418-26 Mfr # 3010X
COMPARE
30 1 Yes
$2,010.00USD / Each
Eagle 4510X Flammable Liquid Safety Cabinet, Self Closing, 2 Doors, 45 Gal EW-09418-27 Mfr # 4510X
COMPARE
45 2 Yes
$2,352.00USD / Each
Eagle 6010X Flammable Liquid Safety Cabinet, Self Closing, 2 Doors, 60 Gal EW-09418-28 Mfr # 6010X
COMPARE
60 2 Yes
$3,706.00USD / Each
Eagle 1992X Flammable Liquid Safety Cabinet, Manual Close, 2 Doors, 90 Gal EW-09418-29 Mfr # 1992X
COMPARE
90 2 No
$3,702.00USD / Each
Eagle 9010X Flammable Liquid Safety Cabinet, Self Closing, 2 Doors, 90 Gal EW-09418-31 Mfr # 9010X
COMPARE
90 2 Yes
$3,832.00USD / Each
Eagle 1945X Flammable Liquid Safety Cabinet, Sliding Self Close, 2 Doors, 45 Gal EW-09418-40 Mfr # 1945X
COMPARE
45 2 Yes
$2,712.00USD / Each
Eagle 1947X Flammable Liquid Safety Cabinet, Manual Close, 2 Doors, 45 Gal EW-09418-70 Mfr # 1947X
COMPARE
45 2 No
$2,747.00USD / Each
Eagle 1962X Flammable Liquid Safety Cabinet, Manual Close, 2 Doors, 60 Gal EW-09418-92 Mfr # 1962X
COMPARE
60 2 No
$2,033.00USD / Each
Showing 1- 12 of 12