Extech Type-K Thermocouple Chisel-Tip Penetration Probe
Ideal for immersion and air temperature measurements
  • Use these standard probes with liquids, gases, and semisolids
  • 6" L stainless steel probe
  • Temperature range: -40 to 800°C (-40 to 1472°F)
  • 1.8 m (6 ft) long coiled cable
Compatible with multimeters and thermometers with type-k mini-connector input.
$60.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Extech 881603 Type-K, Chisel-Tip Penetration Probe, Mini-Connector, 6" L, 0.126" Dia, Grounded EW-95001-54 Mfr # 881603
COMPARE
Type-K, Chisel-Tip Penetration Probe, Mini-Connector, 6" L, 0.126" Dia, Grounded
$60.00USD / Each