Fluke Calibration (formerly Hart Scientific) 1551/1552 Intrinsically Safe Portable Stick Reference Thermometers
The best and most accurate way to replace glass thermometers
  • Built-in probe with a stainless steel sheath ideal for rugged applications
  • Display temperature in Celsius or Fahrenheit
  • ±0.05°C accuracy over full range
  • Temperature trend indicator
  • 200 hour battery life
Whether you’re verifying the accuracy of a temperature sensor or performing a procedure for the pharmaceutical industry, the Fluke Stik thermometer from Hart Scientific gives you a safe, portable, and accurate solution for all of your temperature calibration applications. The probe and digital readout are fixed together, calibrated as a system, and accurate to ±0.1°F (±0.05°C) over the full temperature range with selectable resolution of 0.1, 0.01 or 0.001. The instrument comes with an easy-to-read temperature trend indicator that you configure to meet your application’s stability requirements. The indicator tells you if your temperature is trending up or down and lets you know when your temperature source is stable enough for an accurate measurement. The highly accurate RTD probe is hinged and swivels 90° allowing you to easily view your temperature measurement from the large backlit LCD. The Stik Thermometer operates on three AAA batteries and includes a low-battery indicator and auto-off function that shuts down the thermometer to prevent erroneous measurements due to low battery life.
Available for sale in the United States and Canada only.
ITEM(S) ON SALE
$126.00 - $1,170.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Meter Measurement Type Number Of Probe Channels
Availability Pricing
Fluke Calibration 1551A-9 Handheld Thermometer, Fixed RTD, –50 to 160C, 3/16x9 EW-93202-07 Mfr # 1551A-9
COMPARE
320 RTD 1
$868.50USD / Each
$965.00USD / Each
Fluke Calibration 1551A-12 Handheld Thermometer, Fixed RTD, –50 to 160C, 1/4x12 EW-93202-08 Mfr # 1551A-12
COMPARE
320 RTD 1
$837.00USD / Each
$930.00USD / Each
Fluke Calibration 1552A-12 Handheld Thermometer, Fixed RTD, –80 to 300C, 1/4x12 EW-93202-09 Mfr # 1552A-12
COMPARE
572 RTD 1
$1,170.00USD / Each
Fluke Calibration 1551-CASE Carrying Case, for 1551 / 1552 EW-93202-11 Mfr # 1551-CASE
COMPARE
$126.00USD / Each
$140.00USD / Each