Fluke Calibration 5618B Series - Industrial PRT/RDT probes
5618B Series – Small Diameter Industrial PRT/RTD probes:
For secondary level performance with full ITS-90 calibration, these PRTs are an excellent choice for critical temperature measurements. Featuring a 3.2 mm diameter (1/8 in) sheath, these industrial standards probes have reduced response time without compromising precision. This small diameter 5618A probe works well in many applications where immersion depth is limited.
Available for sale in the United States and Canada only.
$1,770.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
Fluke Calibration 5618B-9-D Secondary PRT Probem 9" x 1/8" OD (S) EW-90449-22 Mfr # 5618B-9-D
COMPARE
932 PRT/RTD
$1,770.00USD / Each
Fluke Calibration 3376977 Industrial PRT Probe, 3.2 mm x 12" Info-Con EW-90450-47 Mfr # 5618B-12-P
COMPARE
932 PRT/RTD
$1,770.00USD / Each
Fluke Calibration 3003331 Industrial PRT Probe, 3.2 mm x 6" , Din EW-90450-49 Mfr # 5618B-6-D
COMPARE
572 PRT/RTD
$1,770.00USD / Each
Fluke Calibration 3376989 Industrial PRT Probe, 3.2 mm x 6" Info-Con EW-90450-52 Mfr # 5618B-6-P
COMPARE
572 PRT/RTD
$1,770.00USD / Each
Fluke Calibration 3376992 Industrial PRT Probe, 3.2 mm x 9" Info-Con EW-90450-56 Mfr # 5618B-9-P
COMPARE
932 PRT/RTD
$1,770.00USD / Each