General Tools Inspection Tools

View and retrieve objects in hard-to-reach areas
  • Telescopic shafts allow extended reach
  • Adjustable mirror
These inspection tools offer telescopic shafts that allow extended reach to inaccessible spaces. Mirrors adjust to any angle for visual examination in trouble areas. Magnets, mirrors 09914-00 and 09914-04, and inspection kit feature a heavy-duty pocket clip.
$17.60 - $34.40USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Adjustable Handle Length Mirror Type Description
Availability Pricing
COMPARE
5-1/2" to 27" Round Round Glass Inspection Mirror, 1.25" Diameter, 5.5-27" Adj. Length
$17.60USD / Each
COMPARE
7-1/4" to 14-1/2" Round 2 1/4" Round Glass Mirror With Cushion grip
$23.80USD / Each
General Tools Rectangular Metal Inspection Mirror, 2"L x 3.75"W, 10.75-26" Adj Length EW-09914-14 Mfr # 5562
COMPARE
10-3/4" to 26" Rectangular Rectangular Metal Inspection Mirror, 2"L x 3.75"W, 10.75-26" Adj Length
$25.50USD / Each
General Tools Round Metal Inspection Mirror, 3.75" Diameter, 9.75-26" Adj. Length EW-09914-16 Mfr # 5563
COMPARE
9-3/4" to 26" Round Round Metal Inspection Mirror, 3.75" Diameter, 9.75-26" Adj. Length
$34.40USD / Each
General Tools Pick-up Magnet, 3.55 lb Capacity, 5 to 25"L Telescoping Handle EW-09914-24 Mfr # 383NX
COMPARE
5" to 25" n/a Pick-up Magnet, 3.55 lb Capacity, 5 to 25"L Telescoping Handle
$17.80USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat