Cole-Parmer® Soil pH Electrode
Soil measurements without the "Suspension effect"
  • Refillable, glass-body electrode allows you to immerse the bulb without immersing the ceramic junction
Our refillable soil electrode has an extra-wide 7.5 mm ceramic junction. Soil electrodes have a maximum temperature range of 212°F
(100°C) and measure 145 mm L x 12 mm dia. Ag/AgCl ref cell.
Use with any meter that accepts a BNC connector, or check out our selection of pH/Ion meters.
ITEM(S) ON SALE
$232.02USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
ATC Element Electrode Body Material Electrode Type
Availability Pricing
COMPARE
Glass Single junction
$232.02USD / Each
$257.80USD / Each