Cole-Parmer® Spex® Freezer/Mill® Cryogenic Grinders
Grind the toughest samples at cryogenic temperatures
  • Reach cryogenic temperatures in minutes
  • Dual grinding and cooling chambers
  • User-friendly touch screen controls
  • 20 user-defined programs for easy quick recall
  • Enhanced safety features
The Freezer/Mill® programmable cryogenic grinder pulverizes through the toughest samples at cryogenic temperatures. Samples are placed in a vial and immersed in a self-contained insulated liquid nitrogen (LN2) tub, reaching cryogenic temperatures in minutes, where they are then pulverized with a magnetically driven impactor. The closed grinding vial impactor prevents sample cross-contamination. The dual grinding and cooling chambers allow you to pre-cool and grind two sets of samples simultaneously.
Grind temperature sensitive samples while maintaining constant temperature, as the sample never leaves the enclosed liquid nitrogen bath. Enhanced safety features include a liquid nitrogen sensor and lid interlock. If the lid is opened while in process, grinding will stop. Auto-fill system is available for automated liquid nitrogen filling.
The large, high-resolution touch screen controls all parameters of the run. Store up to 20 user-defined grinding programs for quick and easy recall. The user-friendly interface allows you to view training videos, user manual, and run history, which can be transferred to a computer using a thumb drive.
Purchase liquid nitrogen, extractor, and sample vials separately.
$11,500.00 - $21,995.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Tubes Held Description
Availability Pricing
COMPARE
0.1 to 5 g 3 Freezer/Mill® Compact Cryogenic Grinder, 0.1 to 5 g; 115 VAC
$11,500.00USD / Each
COMPARE
0.1 to 100 g 4 Freezer/Mill® Large Cryogenic Grinder, 0.1 to 100 g; 115 VAC
$16,900.00USD / Each
COMPARE
0.1 to 100 g 4 Freezer/Mill® Large Cryogenic Grinder with Liquid Nitrogen Autofill, 0.1 to 100 g; 115 VAC
$19,000.00USD / Each
COMPARE
0.1 to 200 g capacity (0.1 to 100 g per chamber) 4 Freezer/Mill® Dual-Chamber High-Capacity Cryogenic Grinder, 0.1 to 200 g; 115 VAC
$21,995.00USD / Each
COMPARE
0.1 to 5 g 3 Freezer/Mill® Compact Cryogenic Grinder, 0.1 to 5 g; 230 VAC
$11,500.00USD / Each
COMPARE
0.1 to 100 g 4 Freezer/Mill® Large Cryogenic Grinder, 0.1 to 100 g; 230 VAC
$16,900.00USD / Each
COMPARE
0.1 to 100 g 4 Freezer/Mill® Large Cryogenic Grinder with Liquid Nitrogen Autofill, 0.1 to 100 g; 230 VAC
$19,000.00USD / Each
COMPARE
0.1 to 200 g capacity (0.1 to 100 g per chamber) 4 Freezer/Mill® Dual-Chamber High-Capacity Cryogenic Grinder, 0.1 to 200 g; 230 VAC
$21,995.00USD / Each
Showing 1- 8 of 8