Cole-Parmer® SamplePrep Mixer/Mill® High-Energy Ball Mills
Easily pulverize hard or brittle samples to analytical fineness in minutes
  • Steel housing protects the clamp mechanism and rugged construction ensures a long life of heavy work.
  • Shock mounted motor enables bench top operation.
  • Equipped with forced-air cooling and safety interlock system for operator protection.
  • Vibration detection switch shuts down the mill in the event of mechanical failure.
  • Electronic timer is factory-set for 100-minute range. Extended Running Time Chip is available for extending timer range to 175 hours (10,500 minutes).
Use these powerful ball mills for mechanical alloying, slurry grinding, blending powders, mixing emulsions, and mechanochemistry. The sample is placed in a grinding vial with one or more balls, then shaken vigorously. The clamp swings back-and-forth 2-1/3” (5.9 cm) with a short lateral “figure-8” movement, producing strong G-forces on the vial to pulverize the toughest samples. Adjustable clamp accepts vials in various materials to effectively grind all sample types and to avoid contamination from specific elements. View time remaining on the digital display. Pause and stop buttons let you pause or stop any time during the process. Order dual-clamp model for high-throughput applications.
These rugged mixer mills come with many safety features. Vibration detection switch shuts down the mill in the event of mechanical failure. Safety interlock and lid latch ensure user safety. Forced air cooling fan prevents overheating. Shock-mounted electric motor cuts back vibration and prolongs motor life. Steel housing and rugged construction ensure a long life of heavy work.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC) Description
Availability Pricing
Cole-Parmer SamplePrep BM-450 Mixer/Mill Dual High-Energy Ball Mill; 115 VAC, 60 Hz EW-04578-10 Mfr # 8000D-115
COMPARE
Mixer/Mill® Dual-Clamp High-Energy Ball Mill; 115 VAC, 60 Hz
Call For Price
Cole-Parmer SamplePrep BM-450 Mixer/Mill Dual High-Energy Ball Mill; 230 VAC, 50 Hz EW-04578-11 Mfr # 8000D-230
COMPARE
1425 230 Mixer/Mill® Dual-Clamp High-Energy Ball Mill; 230 VAC, 50 Hz
Call For Price
Cole-Parmer SamplePrep BM-400 Mixer/Mill® High-Energy Ball Mill; 115 VAC, 60 Hz EW-04578-12 Mfr # 8000M-115
COMPARE
1725 115 Mixer/Mill® Single-Clamp High-Energy Ball Mill; 115 VAC, 60 Hz
Call For Price
Cole-Parmer SamplePrep BM-400 Mixer/Mill® High-Energy Ball Mill; 230 VAC, 50 Hz EW-04578-13 Mfr # 8000M-230
COMPARE
1425 230 Mixer/Mill® Single-Clamp High-Energy Ball Mill; 230 VAC, 50 Hz