Traceable® NSF-Certified Folding Stem Thermometers with Calibration
Fold-up probe to protect from damage
  • NSF-certified—ideal for use in the food industry
  • Rapid-response means readings within 0.9°C in 2 to 3 seconds
  • Use in messy and wet areas—IP65 rated and has antimicrobial coating
  • Foldable stem can be used at any angle
These digital thermometers are ideal for food applications. The silver-impregnated coating is 99% effective, and 100% safe! The stainless steel probe is resistant to both corrosion and chemicals. Fold-up penetration sensor protects both the user and the probe from damage.
Obtain a measurement within 0.9°C in seconds! The wide range covers 99% of routine temperature checks: –40.0 to 250.0°C, with a resolution of 0.1° and ±0.5°C accuracy from –20 to 250°C, ±1.0°C otherwise.
The IP65 rating is ideal for wet areas. Protected against splashes and minor jets of water (like those from a tap), and ideal for kitchen and lab use. A magnetic back lets you stick the thermometer in convenient locations; its compact, foldable size lets you put it in your pocket.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
$116.75 - $128.75USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Probe Length (in)
Availability Pricing
Traceable Folding Stem Thermometer, NSF-Certified; 2.8"L Stainless Steel Probe
Always in Stock
EW-98768-61 Mfr # 6830TR
COMPARE
2 3/4
$116.75USD / Each
Traceable Folding Stem Thermometer, NSF-Certified; 4.5"L Stainless Steel Probe
Always in Stock
EW-98768-62 Mfr # 6831TR
COMPARE
4 1/2
$128.75USD / Each