Traceable® Large-Digit Four-Channel Alarm Timer
Large 3/4"H display—easily view time from across the lab
  • Memory returns display to previously programmed countdown time with touch of a button
  • Extra-loud alarms sound for 1 minute
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
This versatile four-channel laboratory timer is ideal for research laboratories, clinical settings, quality control processes, and industrial applications where precise timing is essential. The large, easy-to-read display and intuitive controls allow for seamless operation, even when wearing gloves. Loud, distinct alarms for each channel ensure clear notifications, reducing the risk of missed steps in critical processes. Designed for flexibility, it can be used on the bench, attached to lab equipment, or taken into the field.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
$42.80USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Number of Channels Timing Capacity
Availability Pricing
Traceable Large-Digit Four-Channel Alarm Timer
Always in Stock
EW-90225-35 Mfr # 5004TR
COMPARE
±0.01% 4 99 hours, 59 minutes, 59 seconds
$42.80USD / Each