CYTIVA Whatman™ Venting Filters

Manufactured under strict clean room conditions and totally biosafe
  • Biotech: sterile vents and exhausts for growth environments
  • Pharmaceutical and diagnostics: vents for liquid vessels
  • Food and beverage: vents for holding, shipping, and dispensing vessels
These filters are ideal for venting and isolation applications to protect vessel contents from external contaminants in biotechnology, pharmaceutical, and food and beverage industries. Filters are also used to protect the environment from contaminants within the vessel. Filters feature a polypropylene housing and a 0.2 µm PTFE membrane allowing easy scale up within the range based on vessel capacity and air flow rate required. Filters are bi-directional and autoclavable.

CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.

ITEM(S) ON SALE
$165.40 - $351.75USD / Pkg of 50

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Whatman 6713-0425 Venting Filter, Disc, 25mm, 50/pk EW-29700-60 Mfr # 6713-0425
COMPARE
Venting Filter, Disc, 25mm, 50/pk
$351.75USD / Pkg of 50
Whatman 6713-1650 Venting Filter, Disc, 50mm, 10/pk EW-29700-62 Mfr # 6713-1650
COMPARE
Venting Filter, Disc, 50mm, 10/pk
$174.36USD / Pkg of 10
$317.40USD / Pkg of 10
Whatman 6713-5036 Venting Filter, Capsule, 500 cm2 EW-29700-64 Mfr # 6713-5036
COMPARE
Venting Filter, Capsule, 500 cm2
$165.40USD / Each
Whatman 6713-1075 Venting Filter, Capsule, 1000 cm2 EW-29700-66 Mfr # 6713-1075
COMPARE
Venting Filter, Capsule, 1000 cm2
$234.95USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat