Conductivity Electrodes
Conductivity Electrodes
Conductivity electrodes work with conductivity meters to measure the electrical conductivity in a solution. The conductivity cell surface is usually platinum, titanium, gold-plated nickel, or graphite. When choosing conductivity cells keep in mind the cell constant, body material, electrode material, ATC element, fitting, cell type, and temperature range.
APPLY FILTERS
filter byclear all
20 RESULTS
SORT BY
Showing 1 - 20 of 20