A&D Weighing AD-1684A Electrostatic Field Meter
Representative image only.
A&D Weighing AD-1684A Electrostatic Field Meter
A&D Weighing | Mfr: AD-1684A| Item: EW-11109-90
Enhance the versatility of your A&D Weighing Balance
$1,846.25USD / Each
$2,110.00USD / Each
$1,846.25USD / Each
$2,110.00USD / Each
Product Description
Key Features
  • Measure the amount of the electrostatic charge — displayed as a bar graph
  • Compatible with A&D GF and GX series balances
  • 1 year warranty
More About this Item
Choose from a wide range of accessories compatible with your A&D Weighing Balance for optimal performance.
This is an electrostatic fieldmeter designed for detecting the existence of static electricity, which is often the cause of inaccurate weighing.
1 Year Warranty
Specifications
  • DescriptionElectrostatic Field Meter