A&D Weighing GXA-26 External IR Switch
Representative image only.
A&D Weighing GXA-26 External IR Switch
A&D Weighing | Mfr: GXA-26| Item: EW-11109-87
Communicate with your A&D balance using compatible output and relay kits
$190.90USD / Each
$230.00USD / Each
$190.90USD / Each
$230.00USD / Each
Product Description
Key Features
  • Compatible with A&D Apollo GF and GX series balances
  • For touchless operation of print or tare
  • 1 year warranty
More About this Item
Operate the balance in the same way as the PRINT key or the RE-ZERO key located on the balance.
1 Year Warranty
Specifications
  • DescriptionExternal IR Switch