Acros Organics 1,4-Dioxane, Extra Dry, Stabilized, 99.8%, 100 mL

Representative image only.

Acros Organics 1,4-Dioxane, Extra Dry, Stabilized, 99.8%, 100 mL

Acros Organics | Mfr: AC326891000| Item: EW-88294-96

Quality and consistency your lab can count on

$146.75USD / Each

$146.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmful
Specifications
  • Concentration99.8%
  • Purity GradeExtra Dry
  • Empirical FormulaC4H8O2
  • ClassReagent Solvents
  • ChemistrySolvents
  • Element/Ion1,4-Dioxane
  • FormLiquid
  • SynonymsDiox
  • Volume (mL)100
  • Formula Weight88.11
  • PackagingAcroSeal™ Glass Bottle
  • DOT InformationDioxane; Class 3, Packing Group II
  • Hazard SymbolHarmful, Highly Flammable
  • CAS Number123-91-1
  • UN NumberUN1165
  • Boiling Point C (° C)101
  • Boiling Point F (° F)213.8
  • Melting Point C (° C)12
  • Melting Point F (° F)53.6
  • Specific Gravity1.034
  • Description1,4-Dioxane, Extra Dry, Stabilized, 99.8%, 100 mL
Documents & Certificates
Reviews
Q&A