Built-in rechargeable battery, and AC power adapter. Draft shield is included only on models with a pan size of 4-3/4" (12cm). (Units with large pans can use an external draft shield sold separately)
Key Features
Internal rechargeable battery for portability and convenience
RS-232 interface allows for communication with a printer
Print function for GLP compliance
Capacity tracker warns of approaching overloads
Security lock-down port
1200 g capacity - robust portable weighing
More About this Item
Cost-Effective Precision Weighing for Everyday Use
Choosing this affordable precision weighing solution doesn't mean sacrificing accuracy or essential features. It provides precise measurements without breaking the budget, making it a cost-effective choice for businesses and laboratories.
Internal Rechargeable Battery for Portability
The inclusion of an internal rechargeable battery adds a level of portability and convenience. Users can take the equipment to various locations without being tethered to a power source, making it suitable for fieldwork, remote sites, and other mobile applications.
Seamless Communication with RS-232 Interface
Communicate with ease. The RS-232 interface connects with a printer or other external devices, streamlining data recording and management. This connectivity option simplifies documentation and data transfer, enhancing workflow efficiency.
Print Function for GLP Compliance
The built-in print function ensures GLP (Good Laboratory Practice) compliance. It simplifies generating and maintaining records, making it easier for users to adhere to regulatory requirements and quality standards.
Capacity Tracker for Overload Warning
A proactive alert system prevents damage to the equipment and ensures accurate measurements, extending the life of the balance and reducing the risk of costly repairs or replacements.
Keeping Equipment Safe
A security lock-down port safeguards valuable precision weighing equipment in shared or high-traffic environments.