Acros Organics Ammonium Hydroxide, 28-30 wt% Solution of NH3 in Water, p.a.; 2.5 L

Representative image only.

Acros Organics Ammonium Hydroxide, 28-30 wt% Solution of NH3 in Water, p.a.; 2.5 L

Acros Organics | Mfr: AC205840025| Item: EW-88220-10

Quality and consistency your lab can count on

$313.75USD / Each

$313.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Corrosive
Specifications
  • Linear FormulaNH4OH
  • FormLiquid
  • Volume (mL)2500
  • Formula Weight35.05
  • CAS Number1336-21-6
  • DescriptionAmmonium hydroxide, 28-30 wt% solution ofNH3 in water p.a. (2.5 L)
Documents & Certificates
Reviews
Q&A