Acros Organics Ammonium Nitrate for Analysis, 99+%, 500 g

Representative image only.

Acros Organics Ammonium Nitrate for Analysis, 99+%, 500 g

Acros Organics | Mfr: AC205865000| Item: EW-88220-15

Quality and consistency your lab can count on

$159.00USD / Each

$159.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

OxidizingIrritant
Specifications
  • Concentration99+%
  • Purity GradeFor analysis
  • Empirical FormulaH4N2O3
  • Linear FormulaNH4NO3
  • ClassReagent Salts
  • ChemistrySolutions
  • Element/IonAmmonium nitrate
  • FormSolid
  • SynonymsNitric acid, ammonium salt; Norway saltpeter
  • Weight (g)500
  • Formula Weight80.04
  • PackagingPlastic bottle
  • DOT InformationAmmonium nitrate; Class 5.1, Packing Group III
  • Hazard SymbolHarmful, Oxidizing
  • CAS Number6484-52-2
  • UN NumberUN1942
  • Boiling Point C (° C)210
  • Melting Point C (° C)169
  • Melting Point F (° F)336.2
  • Specific Gravity1.72
  • DescriptionAmmonium Nitrate for Analysis, 99+%, 500 g
Documents & Certificates
Reviews
Q&A