Acros Organics Benzethonium chloride 97% (100g) CAS 121-54-0

Representative image only.

Acros Organics Benzethonium chloride 97% (100g) CAS 121-54-0

Acros Organics | Mfr: AC105381000| Item: EW-88093-03

Quality and consistency your lab can count on

$109.50USD / Each

$109.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Harmful
Specifications
  • Empirical FormulaC27H42ClNO2
  • ClassReagent Salts
  • ChemistrySolutions
  • Synonyms(Diisobutylphenoxyethoxyethyl)dimethylbenzylammonium chloride, Hyamine 162
  • FormSolid
  • Weight (g)100
  • Formula Weight448.08
  • DOT InformationEnvironmentally Hazardous Substance, solid, n.o.s.; Class 9, Packing Group III
  • Hazard SymbolHarmful
  • CAS Number121540
  • UN NumberUN3077
  • DescriptionBenzethonium chloride 97% (100g) CAS 121-54-0
Documents & Certificates
Reviews
Q&A