Acros Organics Dichloromethane, stabilized, spectrophotometric grade 99.8% (500 mL)

Representative image only.

Acros Organics Dichloromethane, stabilized, spectrophotometric grade 99.8% (500 mL)

Acros Organics | Mfr: AC167775000| Item: EW-88177-99

Quality and consistency your lab can count on

$87.20USD / Each

$87.20USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Harmful
Specifications
  • Purity GradeSpectrophotometric
  • Empirical FormulaCH2Cl2
  • Linear FormulaCH2Cl2
  • ClassReagent Solvents
  • ChemistrySolutions
  • FormLiquid
  • SynonymsMethylene chloride
  • Volume (mL)500
  • Formula Weight84.93
  • PackagingGlass bottle
  • DOT InformationDichloromethane; Class 6.1, Packing Group III
  • Hazard SymbolHarmful
  • UN NumberUN1593
  • CAS Number75-09-2
  • DescriptionDichloromethane, stabilized, spectrophotometric grade 99.8% (500 mL)
Documents & Certificates
Reviews
Q&A