Acros Organics Ether anhydrous, certified ACS (500 mL) CAS 60-29-7

Representative image only.

Acros Organics Ether anhydrous, certified ACS (500 mL) CAS 60-29-7

Acros Organics | Mfr: AC615085000| Item: EW-88366-13

Quality and consistency your lab can count on

$128.25USD / Each

$128.25USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmful
Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaC4H10O
  • Linear Formula(CH3CH2)2O
  • ClassReagent Solvents
  • ChemistryAnalytical Reagents;Solutions
  • FormLiquid
  • SynonymsDiethyl ether, Ethyl ether
  • Volume (mL)500
  • Formula Weight74.12
  • DOT InformationDiethyl Ether; Class 3, Packing Group I
  • Hazard SymbolExtremely Flammable, Harmful
  • UN NumberUN1155
  • CAS Number60-29-7
  • DescriptionEther anhydrous, certified ACS (500 mL) CAS 60-29-7
Documents & Certificates
Reviews
Q&A