Acros Organics Ethylenediaminetetraacetic acid (EDTA) 99% (1kg)

Representative image only.

Acros Organics Ethylenediaminetetraacetic acid (EDTA) 99% (1kg)

Acros Organics | Mfr: AC118430010| Item: EW-88113-90

Quality and consistency your lab can count on

$222.25USD / Each

$222.25USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Irritant
Specifications
  • Empirical FormulaC10H16N2O8
  • Linear Formula(HO2CCH2)2NCH2CH2N(CH2CO2H)2
  • ClassBuffers
  • Synonyms(Ethylenedinitrilo)tetraacetic acid disodium salt, Edetic acid disodium salt, EDTA disodium salt
  • FormPowder
  • Weight (g)1000
  • Formula Weight292.23
  • DOT InformationNot regulated by DOT
  • Hazard SymbolIrritant
  • CAS Number60-00-4
  • DescriptionEthylenediaminetetraacetic acid (EDTA) 99% (1kg)
Documents & Certificates
Reviews
Q&A