Acros Organics Hexamethylenetetramine, contains an anticaking agent 98.5% (2.5kg)

Representative image only.

Acros Organics Hexamethylenetetramine, contains an anticaking agent 98.5% (2.5kg)

Acros Organics | Mfr: AC232890025| Item: EW-88247-81

Quality and consistency your lab can count on

$229.50USD / Each

$229.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

FlammableHarmful
Specifications
  • Empirical FormulaC6H12N4
  • ClassSolids
  • ChemistrySolutions
  • SynonymsHexamine; HMTA; Methenamine
  • FormSolid
  • Weight (g)2500
  • Formula Weight140.19
  • PackagingPlastic bottle
  • DOT InformationHexamethylenetetramine; Class 4.1, Packing Group III
  • Hazard SymbolHarmful, Highly Flammable
  • UN NumberUN1328
  • CAS Number100-97-0
  • DescriptionHexamethylenetetramine, contains an anticaking agent 98.5% (2.5kg)
Documents & Certificates
Reviews
Q&A