Acros Organics Polysorbate 80; 1 L

Representative image only.

Acros Organics Polysorbate 80; 1 L

Acros Organics | Mfr: AC278630010| Item: EW-88265-49

Quality and consistency your lab can count on

$239.50USD / Each

$239.50USD / Each

Buy It Together

Current Item

Acros Organics Polysorbate 80; 1 L
Acros Organics Polysorbate 80; 1 L
8826549

$239.50USD / Each
+ =
Total Price 735.00 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • ClassGeneral Solutions
  • ChemistrySolutions
  • SynonymsPolyoxyethylene(20)sorbitan monooleate
  • FormLiquid
  • Volume (mL)1000
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • CAS Number9005-65-6
  • DescriptionTween 80 (1l) CAS 9005-65-6 (Polyoxyethylene(20)sorbitan monooleate)
Documents & Certificates
Reviews
Q&A