Acros Organics Toluidine Blue O, Basic Blue 17; 25 g

Representative image only.

Acros Organics Toluidine Blue O, Basic Blue 17; 25 g

Acros Organics | Mfr: AC348600250| Item: EW-88318-49

Quality and consistency your lab can count on

$280.00USD / Each

$280.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Solubility&dot-ldr;To pass test Loss on Drying at 105°C&dot-ldr;≤10% E1%1cm&dot-ldr;≥700L g−1cm−1 Lambda Max. (λ)&dot-ldr;628nm±2nm

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Empirical FormulaC15H16ClN3S
  • ClassSolids
  • ChemistrySolutions
  • SynonymsBasic Blue 17, C.I. 52040, Tolonium chloride
  • FormSolid
  • Weight (g)25
  • Formula Weight305.82
  • DOT InformationNot regulated by DOT
  • Hazard SymbolFlammable, Harmful
  • CAS Number92-31-9
  • DescriptionAcros Organics Toluidine Blue O, Basic Blue 17; 25 g
Documents & Certificates
Reviews
Q&A