Ashcroft 1009 Glycerin-Filled SS Vacuum Gauge, 3.5" Dial, 1/4" NPT(M) Lower Connection, 30" Hg to 0 psi
Representative image only.
Ashcroft 1009 Glycerin-Filled SS Vacuum Gauge, 3.5" Dial, 1/4" NPT(M) Lower Connection, 30" Hg to 0 psi
Ashcroft | Mfr: 351009SWL 02L 30 INHG VAC| Item: EW-68022-33
Liquid-filled to dampen vibration
$105.80USD / Each
$132.25USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$105.80USD / Each
$132.25USD / Each
Buy It Together
Current Item
Ashcroft 1009 Glycerin-Filled SS Vacuum Gauge, 3.5" Dial, 1/4" NPT(M) Lower Connection, 30" Hg to 0 psi
Ashcroft 1009 Glycerin-Filled SS Vacuum Gauge, 3.5" Dial, 1/4" NPT(M) Lower Connection, 30" Hg to 0 psi
6802233
$105.80USD / Each
$132.25USD / Each
+ +
Ashcroft 4.5" Industrial Process Gauge 30 to 0" Hg
6833700
$109.80USD / Each
$137.25USD / Each
=
Total Price 395.35 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Meets ASME B40.100 specifications
  • Corrosion resistant SS case
  • Dry gauges are field fillable
  • PowerFlex™ movement provides superior resistance to shock, vibration and pulsation
  • True Zero™ reduces reading errors by using “zero box” instead of conventional dial pins
  • MSL helium leak tested to 1X10-6 scc/s
  • Ventable fill plug
  • 1 year warranty
More About this Item
The Ashcroft® type 1009 Industrial Pressure Gauge is ideal for resisting chemical, weather or corrosion effects. Providing durability, reliability and performance, it is well-suited for aggressive process media applications. A good choice for satisfying many application and installation requirements. Available in 2.5" and 3.5" dial sizes, Ashcroft Duralife pressure gauges are liquid fillable and field convertible for panel mounting. Both zero and span adjustments are standard.
1 Year Warranty
Specifications
  • AccuracyASME B40.100, Grade 1A, +1% of span (Dry), +1.5% of span (Liquid-filled)
  • Dial Size (in)3.5
  • Dial Size (mm)100
  • Max Vacuum (" Hg)30
  • Min Vacuum (" Hg)0
  • Liquid FillGlycerin
  • Wetted Materials316 stainless steel
  • Case304 stainless steel
  • Lens MaterialPolycarbonate
  • Process Connection1/4" NPT(M), lower
  • Unit Of Measurement" Hg
  • DisplayAnalog dial
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionGlycerin-Filled Stainless Steel Vacuum Gauge, 3.5" Dial, 1/4" NPT(M) Lower Connection, 30" Hg to 0 psi