Autovial AV125EAQU Whatman Syringeless Filters, PVDF; 0.20µ; 50/box
Representative image only.
Autovial AV125EAQU Whatman Syringeless Filters, PVDF; 0.20µ; 50/box
Autovial | Mfr: AV125EAQU| Item: UX-29703-20
Direct instrument injection
$445.75USD / Box of 50
$445.75USD / Box of 50
Product Description
Key Features
  • Integral filter
  • Built-in air purge
  • Recessed slip-luer tip
  • Use PVDF filters (<60% organic) with low nonspecific protein binding for samples in aqueous solution and solvent or aqueous/organic solvents
More About this Item
Autovial syringeless filters offer maximum convenience. Simply fill the 12-mL graduated polypropylene filter barrel and press the plunger to push sample through. Select your filter based on membrane material and its compatibility to your sample (see application selection guide below). Membrane materials are nylon, PTFE (PTFE), polyvinylidene fluoride (PVDF), glass microfiber (GMF), and mixed ester (ME).
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • Membrane MaterialPolyvinylidene fluoride (PVDF)
  • Pore Size (µm)0.2
  • Housing MaterialPolypropylene (PP)
  • DescriptionWhatman Syringeless Filters, PVDF; 0.20µ; 50/box