Whatman Autovial Syringeless Filters
Direct instrument injection
  • Integral filter
  • Built-in air purge
  • Recessed slip-luer tip
Autovial syringeless filters offer maximum convenience. Simply fill the 12-mL graduated polypropylene filter barrel and press the plunger to push sample through. Select your filter based on membrane material and its compatibility to your sample (see application selection guide below). Membrane materials are nylon, PTFE (PTFE), polyvinylidene fluoride (PVDF), glass microfiber (GMF), and mixed ester (ME).
$445.75 - $474.10USD / Box of 50
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Material Pore Size (µm)
Availability Pricing
Autovial AV125EORG Whatman Syringeless Filters, PTFE; 0.20µ; 50/box EW-29703-10 Mfr # AV125EORG
COMPARE
Polytetrafluoroethylene (PTFE) 0.2
$473.05USD / Box of 50
Autovial AV125UORG Whatman Syringeless Filters, PTFE; 0.45µ; 50/box EW-29703-12 Mfr # AV125UORG
COMPARE
Polytetrafluoroethylene (PTFE) 0.45
$457.80USD / Box of 50
Autovial AV125EAQU Whatman Syringeless Filters, PVDF; 0.20µ; 50/box EW-29703-20 Mfr # AV125EAQU
COMPARE
Polyvinylidene fluoride (PVDF) 0.2
$445.75USD / Box of 50
Autovial AV125UGMF Whatman Syringeless Filters, GMF; 0.45µ; 50/box EW-29703-30 Mfr # AV125UGMF
COMPARE
Glass Microfiber (GMF) 0.45
$474.10USD / Box of 50