Aven Tools Titanium Precision Smooth Round-Tapered Tip 2A Tweezers, 4.75" L; 1/Pk
Representative image only.
Aven Tools Titanium Precision Smooth Round-Tapered Tip 2A Tweezers, 4.75" L; 1/Pk
Aven Tools | Mfr: 18049TT| Item: UX-07289-26
$61.50USD / Each
$61.50USD / Each
Product Description
More About this Item
Titanium tweezers are excellent for the silicon chip industry, for handling microscopic particles, and when working in a magnetic field as they are non-magnetic, lighweight, and strong. They do not shed particles like stainless steel tweezers and are biocompatible. Temperature resistance is up to 800°F(426°C).
Specifications
  • MaterialTitanium
  • SterileNo
  • DescriptionTitanium Precision Smooth Round-Tapered Tip 2A Tweezers, 4.75" L; 1/Pk