Precision Tweezers, Titanium
Titanium tweezers are excellent for the silicon chip industry, for handling microscopic particles, and when working in a magnetic field as they are non-magnetic, lighweight, and strong. They do not shed particles like stainless steel tweezers and are biocompatible. Temperature resistance is up to 800°F(426°C).
$61.30 - $62.40USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Sterile Description
Availability Pricing
Aven Tools Titanium Precision Smooth Pointed Tip AA Tweezers, 4.75" L; 1/Pk EW-07289-20 Mfr # 18013TT
COMPARE
Titanium No Titanium Precision Smooth Pointed Tip AA Tweezers, 4.75" L; 1/Pk
$61.30USD / Each
Aven Tools Titanium Precision Smooth Pointed Tip OO Tweezers, 4.75" L; 1/Pk EW-07289-22 Mfr # 18032TT
COMPARE
Titanium No Titanium Precision Smooth Pointed Tip OO Tweezers, 4.75" L; 1/Pk
$61.30USD / Each
Aven Tools Titanium Precision Smooth Round-Tapered Tip 2A Tweezers, 4.75" L; 1/Pk EW-07289-26 Mfr # 18049TT
COMPARE
Titanium No Titanium Precision Smooth Round-Tapered Tip 2A Tweezers, 4.75" L; 1/Pk
$61.50USD / Each
Aven Tools Titanium Precision Smooth Ultra-Fine Tip 3C Tweezers, 4.75" L; 1/Pk
Always in Stock
EW-07289-30 Mfr # 18056TT
COMPARE
Titanium No Titanium Precision Smooth Ultra-Fine Tip 3C Tweezers, 4.75" L; 1/Pk
$61.50USD / Each
Aven Tools Titanium Precision Smooth Ultra-Fine Tip 5 Tweezers, 4.25" L; 1/Pk EW-07289-32 Mfr # 18062TT
COMPARE
Titanium No Titanium Precision Smooth Ultra-Fine Tip 5 Tweezers, 4.25" L; 1/Pk
$62.40USD / Each
Aven Tools Titanium Precision Smooth Ultra-Fine Bent Tip 6 Tweezers, 4.25" L; 1/Pk EW-07289-34 Mfr # 18069TT
COMPARE
Titanium No Titanium Precision Smooth Ultra-Fine Bent Tip 6 Tweezers, 4.25" L; 1/Pk
$61.50USD / Each
Aven Tools Titanium Precision Smooth Fine Curve Tip 7 Tweezers, 4.25" L; 1/Pk EW-07289-36 Mfr # 18072TT
COMPARE
Titanium No Titanium Precision Smooth Fine Curve Tip 7 Tweezers, 4.25" L; 1/Pk
$62.40USD / Each
Showing 1- 7 of 7