Bacharach 19-8025 Carbon dioxide incubator monitor, 0-50%, 110/240 VAC
Representative image only.
Bacharach Mfr # 19-8025 – Item # EW-81973-10
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Powerone NiCd rechargeable battery (included)
  • DescriptionCarbon dioxide incubator monitor, 0-50%, 110/240 VAC
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
CO2 Monitor is a compact and lightweight detection device that immediately responds to excessive levels of CO2—ideal for personal monitoring. Meter features a maintenance-free infrared sensor and audible/visible alarms that respond intermittently to CO2 concentrations above 5000 ppm, and continuously to concentrations above 10,000 ppm.
CO2 Incubator Monitor provides fast, accurate CO2 level monitoring. Features a powerful internal pump for extracting samples; two sampling modes, sample draw and diffusion; integral fan; and maintenance-free dual wavelength pressure and temperature compensated sensors. Keep records of your CO2 levels with the built-in datalogging feature. Choose to log your readings continuously or with the manual snapshot mode. Nonvolatile memory holds approximately 100 snapshot readings and 800 sets of continuous datalogged readings. Download stored readings to your PC for storage or analysis via the infrared data link. Alarm features a variable pitched tone to indicate CO2 levels.