TPI CO and Flue Analyzers
Choose from a complete line of flue gas and CO analyzers
  • When selecting oil as fuel be sure to use the optional oil filter TP773 or reading could become erratic. You must use the TP715K3 kit which includes the TP784 regulator when working on oil and gas pipeline diesel compression stations The TP784 flow regulator must be used in any application where high velocity pressure is present
Choose from a complete line of flue gas and CO analyzers. All units measure CO and depending on the model, you can additionally measure O2, temperature, pressure, and NO. Calculate CO 2, gross of net efficiency, excess air and NOX. All models include flue probe and carrying case. Select the model that is right for you.
$418.00 - $607.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max NO2 Range (ppm) Max NO Range (ppm) Max SO2 Range (ppm)
Availability Pricing
COMPARE
$418.00USD / Each
COMPARE
$552.00USD / Each
COMPARE
$607.00USD / Each