Chemetrics K-3510 Colorimetric Copper Test Kit

Representative image only.

Chemetrics K-3510 Colorimetric Copper Test Kit

Chemetrics | Mfr: K-3510| Item: EW-00290-50

CHEMets test kits are portable and self-contained

$103.35USD / Each

$103.35USD / Each

Buy It Together

Current Item

Chemetrics K-3510 Colorimetric Copper Test Kit
Chemetrics K-3510 Colorimetric Copper Test Kit
0029050

$103.35USD / Each
+ + +
LaMotte Insta-Test Ammonia Test Strips, 0 to 6 ppm, 25/Pk
9953264

$38.25USD / Pkg of 25
=
Total Price 286.35 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Vacuum sealed reagents and handy comparator chart for quick, visual results

More About this Item

CHEMETS colorimetric test kits let you visually compare the color of your sample to standards using a comparator. Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely.

Specifications
  • TestCopper, total
  • Measurement Points0 to 1 ppm; 1 to 10 ppm
  • Reagent PlatformBathocuproine
  • Tests Per Kit30
  • DescriptionColorimetric Copper Test Kit
Reviews
Q&A