Cole-Parmer Ascorbic Acid, ACS Grade, 500 g

Representative image only.

Cole-Parmer Ascorbic Acid, ACS Grade, 500 g

Cole-Parmer | Item: EW-88403-08

Reagents and solutions to meet your lab and research needs

$459.00USD / Each

$459.00USD / Each
Product Description

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS Reagent
  • Empirical FormulaC6H8O6
  • ClassReagent Acids
  • FormSolid
  • Weight (g)500
  • Formula Weight176.12
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number50-81-7
  • DescriptionAscorbic Acid, ACS Grade, 500 g
Reviews
Q&A