Cole-Parmer Autoradiography Cassette, Aluminum, 8 x 10"
Representative image only.
Cole-Parmer Autoradiography Cassette, Aluminum, 8 x 10"
Cole-Parmer | Item: EW-21700-35
Get the results you need affordably
$378.00USD / Each
$378.00USD / Each
Product Description
Key Features
  • Constructed from lightweight aluminum and nylon
  • Special curved door gives exceptional screen-film contact
More About this Item
Aluminum Cassettes have a push-button front for easy use. The lightweight cassettes feature a curved door with a nylon-covered foam cushion to provide exceptional screen contact.
Specifications
  • MaterialAluminum and Nylon
  • Length (InchShort)8
  • Width (InchShort)10
  • DescriptionAutoradiography Cassette, Aluminum, 8 x 10"