Cole-Parmer Column Clamp, Large, Stainless Steel, 2.52" to 5.51" Grip Range
Representative image only.
Cole-Parmer Column Clamp, Large, Stainless Steel, 2.52" to 5.51" Grip Range
Cole-Parmer | Item: EW-08064-10
Provide support for your specific laboratory equipment
$105.00USD / Each
$105.00USD / Each
Product Description
Key Features
  • Durable stainless steel clamps offer chemical resistance and increased strength
  • 1 year warranty
More About this Item
Ideal for large cylindrical glassware and similar labware. The large, flat thumbscrew/worm-drive provides a firm hold. NOTE: An additional connector must be purchased in order to attach clamps to frames or other apparatus.
1 Year Warranty
Specifications
  • Clamp MaterialStainless Steel
  • Clamp TypeEquipment, Column
  • Min Grip Range (InchShort)2.52
  • Max Grip Range (InchShort)5.51
  • Length (in)13.3
  • DescriptionColumn Clamp, Large, Stainless Steel, 2.52" to 5.51" Grip Range