Cole-Parmer CP Agarose LE 500g

Representative image only.

Cole-Parmer CP Agarose LE 500g

Cole-Parmer | Item: UX-00141-03

Increase electrophoretic mobility—low EEO

$521.00USD / Each
PART OF A COMPLETE SYSTEM – GET EVERYTHING YOU NEED
Mini Dual Gel Units. Small format is ideal for mini SDS-PAGE and mini agarose gel separations, plus it reduces the use of expensive reagents.
Product Description

Key Features

  • Enhance resolution and clarity with this high purity powder
  • RNase, DNase, and Protease free
  • Obtain faster run times

More About this Item

Cole-Parmer® Agarose LE is a highly purified powder, manufactured without organic solvents, to work on protein and nucleic acid analysis from 150 bp to 25 kbp. The low EEO helps with increased electrophoretic mobility to obtain increased resolution and shorter run times. The powder dissolves completely and forms an ultra-transparent gel. Use for both preparative and analytical nucleic acid electrophoresis, protein electrophoresis, and various blotting procedures.

Specifications
  • Media Name (No.)Agarose
  • Capacity (g)500
  • DescriptionAgarose LE; 500g
Reviews
Q&A