Cole-Parmer® Agarose LE Powder
Increase electrophoretic mobility—low EEO
  • Enhance resolution and clarity with this high purity powder
  • RNase, DNase, and Protease free
  • Obtain faster run times
Cole-Parmer® Agarose LE is a highly purified powder, manufactured without organic solvents, to work on protein and nucleic acid analysis from 150 bp to 25 kbp. The low EEO helps with increased electrophoretic mobility to obtain increased resolution and shorter run times. The powder dissolves completely and forms an ultra-transparent gel. Use for both preparative and analytical nucleic acid electrophoresis, protein electrophoresis, and various blotting procedures.
$62.10 - $521.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Description
Availability Pricing
COMPARE
25 Agarose LE; 25g
$62.10USD / Each
COMPARE
100 Agarose LE; 100g
$149.75USD / Each
COMPARE
500 Agarose LE; 500g
$521.00USD / Each