Cole-Parmer Economical Bowl, 204 cu Stainless Steel, 1.5 Qt Capacity
Representative image only.
Cole-Parmer Economical Bowl, 204 cu Stainless Steel, 1.5 Qt Capacity
Cole-Parmer | Item: EW-07306-03
204Cu stainless steel for corrosion resistance at a lower cost
$21.40USD / Each
BUY MORE & SAVE
  • 1-5$21.40 USD / Each
  • 6-29$19.26 USD / Each
  • 30-119$18.19 USD / Each
  • 120+$17.12 USD / Each
$21.40USD / Each
BUY MORE & SAVE
  • 1-5$21.40 USD / Each
  • 6-29$19.26 USD / Each
  • 30-119$18.19 USD / Each
  • 120+$17.12 USD / Each
Product Description
Key Features
  • Polished to a mirror shine for easy cleaning and nonstick surface
  • Use for high-temperature applications to 1400°F (760°C)
  • 1 year warranty
1 Year WarrantyNSF international certified
Specifications
  • Capacity (L)1.4
  • Capacity (Qt)1.5
  • Material204 Cu Stainless Steel
  • Overall Height (in)2 3/4
  • Overall Height (cm)7
  • Outside Diameter (OD) (in)7 3/4
  • Outside Diameter (OD) (cm)19.7
  • DescriptionEconomical Bowl, 204 cu Stainless Steel, 1.5 Qt Capacity