Cole-Parmer Economical Bowl, 204 cu Stainless Steel, 3/4 Qt Capacity
Representative image only.
Cole-Parmer Economical Bowl, 204 cu Stainless Steel, 3/4 Qt Capacity
Cole-Parmer | Item: EW-07306-01
204Cu stainless steel for corrosion resistance at a lower cost
$13.10USD / Each
BUY MORE & SAVE
  • 1-5$13.10 USD / Each
  • 6-29$11.79 USD / Each
  • 30-119$11.14 USD / Each
  • 120+$10.48 USD / Each
$13.10USD / Each
BUY MORE & SAVE
  • 1-5$13.10 USD / Each
  • 6-29$11.79 USD / Each
  • 30-119$11.14 USD / Each
  • 120+$10.48 USD / Each
Product Description
Key Features
  • Polished to a mirror shine for easy cleaning and nonstick surface
  • Use for high-temperature applications to 1400°F (760°C)
  • 1 year warranty
1 Year WarrantyNSF international certified
Specifications
  • Capacity (L)0.7
  • Capacity (Qt)0.75
  • Material204 Cu Stainless Steel
  • Overall Height (in)2
  • Overall Height (cm)5
  • Outside Diameter (OD) (in)6 1/2
  • Outside Diameter (OD) (cm)16.5
  • DescriptionEconomical Bowl, 204 cu Stainless Steel, 3/4 Qt Capacity