Cole-Parmer EDTA Titrant, 0.100 Molar (M/10), 4 L

Representative image only.

Cole-Parmer EDTA Titrant, 0.100 Molar (M/10), 4 L

Cole-Parmer | Item: EW-88400-26

Quality and consistency your lab can count on

$243.50USD / Each

$243.50USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Normality0.1
  • Empirical FormulaC10H14N2O8Na2•2H2O
  • ClassTitrants
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight372.24
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7732-18-5, 6381-92-6
  • Shelf Life (months)24
  • DescriptionEDTA Titrant, 0.100 Molar (M/10), 4 L
Reviews
Q&A