Cole-Parmer Ethylene Glycol, Reagent Grade; 1 L

Representative image only.

Cole-Parmer Ethylene Glycol, Reagent Grade; 1 L

Cole-Parmer | Item: EW-88405-52

Quality and consistency your lab can count on

$77.80USD / Each

$77.80USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

HarmfulToxic
Specifications
  • Purity GradeReagent
  • Empirical FormulaHOCH2CH2OH
  • ClassReagent Solvents
  • ChemistrySolvents
  • Element/IonEthylene Glycol
  • Synonyms1,2-Ethanediol
  • FormLiquid
  • Volume (mL)1000
  • Formula Weight62.06
  • PackagingAmber Glass Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number107-21-1
  • Shelf Life (days)60
  • Boiling Point C (° C)197
  • Melting Point C (° C)-11
  • DescriptionEthylene Glycol, Reagent Grade; 1 L
Reviews
Q&A