Cole-Parmer Filter for Comfort-Grip Motorized Pipette Controller, 0.45 um, 5/pk
Representative image only.
Cole-Parmer Filter for Comfort-Grip Motorized Pipette Controller, 0.45 um, 5/pk
Cole-Parmer | Item: EW-25200-89
Optimize performance of your Cole-Parmer pipettes
$51.80USD / Pkg of 5
$51.80USD / Pkg of 5
Buy It Together
Current Item
Cole-Parmer Filter for Comfort-Grip Motorized Pipette Controller, 0.45 um, 5/pk
Cole-Parmer Filter for Comfort-Grip Motorized Pipette Controller, 0.45 um, 5/pk
2520089
$51.80USD / Pkg of 5
+ =
Total Price 464.80 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Keep your pipette running smoothly with these filters and stands
  • 1 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories to ensure top performance of your pipette controller.
Chemical-resistant PTFE filters help prevent volatile fumes and accidental liquid overfill from corroding your pipette controller's internal components.
0.45 µm pore size helps prevent against most fumes and fluids where bacterial contamination is not a large concern.
1 Year Warranty
Specifications
  • Compatible With25200-15
  • DescriptionFilter for Comfort-Grip Motorized Pipette Controller, 0.45 um, 5/pk