Cole-Parmer Flow-Through Cell, Quartz, 1.8 mL

Representative image only.

Cole-Parmer Flow-Through Cell, Quartz, 1.8 mL

Cole-Parmer | Item: EW-83070-48

Optimize performance of your Cole-Parmer® spectrophotometer

$1,010.00USD / Each

$1,010.00USD / Each

Buy It Together

Current Item

Cole-Parmer Flow-Through Cell, Quartz, 1.8 mL
Cole-Parmer Flow-Through Cell, Quartz, 1.8 mL
8307048

$1,010.00USD / Each
+
Cole-Parmer SP-800-UV Double-Beam Spectrophotometer with Variable Bandwidth; 115 VAC
8307004

$9,832.50USD / Each
$13,110.00USD / Each
=
Total Price 10842.50 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Choose accessories compatible with your double-beam spectrophotometers
  • 1 year warranty

More About this Item

Expand the capabilities of your spectrophotometer with these cell, cell holder, changer, sipper, and peltier options.

1 Year Warranty
Specifications
  • DescriptionFlow-Through Cell, Quartz, 1.8 mL
  • Warranty1 year
Reviews
Q&A