Cole-Parmer Heating Block, Triple Wide, Aluminum, Blank with Thermometer Hole
Representative image only.
Cole-Parmer Heating Block, Triple Wide, Aluminum, Blank with Thermometer Hole
Cole-Parmer | Item: EW-36620-60
Interchangeable heating blocks offer excellent thermal conductivity
$628.00USD / Each
$628.00USD / Each
Buy It Together
Current Item
Cole-Parmer Heating Block, Triple Wide, Aluminum, Blank with Thermometer Hole
Cole-Parmer Heating Block, Triple Wide, Aluminum, Blank with Thermometer Hole
3662060
$628.00USD / Each
+ + +
Cole-Parmer Two Block Heater Cover
3661096
$179.00USD / Each
+
Cole-Parmer Replacement Gas Chamber Sealing Pad
9995007
$23.60USD / Each
+ + + + + =
Total Price 7100.10 ADD 10 ITEMS TO CART
Product Description
Key Features
  • Versatile—interchangeable heating blocks accommodate various tubes and plates
  • Block heaters sold separately
  • Triple wide—measures 95 x 225 x 50 mm
  • 1 year warranty
More About this Item
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Depending on the combination of blocks selected, plastic spacers (36620-67) can be used to fill a half-width recess, ensuring optimal thermal performance. Most blocks come with a separate hole to accept a thermometer.
Order a flexi-block to accommodate multiple size tubes, vials, or cuvettes at once. Unskirted PCR plates can be heated in either size flexi-block. Skirted microtiter plates, deep-well plates, and PCR plates can only be incubated in the triple-width version.
1 Year Warranty
Specifications
  • Number Of Blocks3
  • Block Number Of Openings0
  • Block MaterialAluminum
  • DescriptionHeating Block, Triple Wide, Aluminum, Blank with Thermometer Hole